Learn More
Fisherbrand™ Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
 View more versions of this product
Catalog No. 14130W
Only null left
Description
Description
Available in a variety of sizes ranging from 000 through 16
  • Solid rubber construction
Specifications
Chemical Identifiers
Black
Natural Rubber
Solid Stopper
General purpose use
Specifications
Solid Stopper
8.3 cm
3.9 cm
Natural Rubber
General purpose use
10.3 cm
15
15
Black
Product Suggestions
Product Suggestions