missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, 22.2% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company R78000004A
| Quantity | 4 L |
|---|---|
| Packaging | Natural Poly Bottle |
Identifiants chimiques
| 7757-82-6 , 7732-18-5 | |
| 142.036 | |
| sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
| CHEBI:32149 | |
| [O-]S(=O)(=O)[O-].[Na+].[Na+] |
| Na2O4S | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| 24436 | |
| disodium;sulfate |
Spécifications
| Sodium Sulfate | |
| 80.0, 17.81 | |
| Na2O4S | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium;sulfate | |
| 24436 | |
| Natural Poly Bottle |