missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, Strong R, Ricca Chemical
Spécifications
Spécifications
| Color | Colorless |
| CAS Min % | 69.0, 29.41 |
| CAS Max % | 71.0, 30.62 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | >13 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium Hydroxide Solution, Strong |