missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 25% (w/v) (250 g/L), Ricca Chemical
$114.78 - $1402.41
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 40.00 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7273-16
|
Ricca Chemical Company
727316 |
500 mL | Natural Poly Bottle |
Each for $114.78
|
|
||||
|
7273-32
|
Ricca Chemical Company
727332 |
1 L | Natural Poly Bottle |
Each for $178.19
|
|
||||
|
7273-1
|
Ricca Chemical Company
72731 |
4 L | Natural Poly Bottle |
Each for $505.38
|
|
||||
|
7273-5
|
Ricca Chemical Company
72735 |
20 L | Cubitainer |
Each for $1,402.41
|
|
||||
Chemical Identifiers
| 1310-73-2 | |
| 40.00 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium hydroxide |
Specifications
| 1310-73-2,7732-18-5 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium hydroxide | |
| 14798 | |
| 24.94 to 25.06% | |
| 25% (w/v) Aqueous Solution (250 g/L) | |
| Sodium Hydroxide Solution |
| HNaO | |
| 500 mL | |
| [OH-].[Na+] | |
| 40.00 | |
| CHEBI:32145 | |
| Laboratory | |
| Natural Poly Bottle |