missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 25% (w/v) (250 g/L), Ricca Chemical

Supplier:  Ricca Chemical Company 72735

 View more versions of this product

Catalog No. 7273-5


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
HNaO
20 L
[OH-].[Na+]
40.00
CHEBI:32145
Laboratory
Cubitainer
1310-73-2,7732-18-5
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
14798
24.94 to 25.06%
25% (w/v) Aqueous Solution (250 g/L)

Chemical Identifiers

1310-73-2
40.00
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium hydroxide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions