missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 25% (w/v) (250 g/L), Ricca Chemical
Specifications
Specifications
| Quantity | 500 mL |
| Percent Purity | 24.94 to 25.06% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 25% (w/v) Aqueous Solution (250 g/L) |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide Solution |