missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 20% (w/w), Ricca Chemical

Numéro de catalogue. 72711
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
72711 4 L Natural Poly Bottle
72715 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 72711 Fournisseur Ricca Chemical Company Code fournisseur 72711
Il en reste null

Identifiants chimiques

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

CAS Min % 66.0, 32.16
CAS Max % 69.0, 33.48
Percent Purity 20%
Grade Laboratory
Packaging Natural Poly Bottle
pH >13
Quantity 4 L
Physical Form ear colorless liquid
Chemical Name or Material Sodium Hydroxide