Learn More
Sodium hydroxide, 10N aq. soln.
Description
Sodium hydroxide is used to neutralize acids and make sodium salts. Sodium hydroxide is used to manufacture soaps, rayon, paper, explosives, dyestuffs, and petroleum products. It is also used in processing cotton fabric, laundering and bleaching, metal cleaning and processing, oxide coating, electroplating, and electrolytic extracting. It is commonly present in commercial drain and oven cleaners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Colorless |
| Linear Formula | NaOH |
| UN Number | UN1824 |
| Concentration | 10N aq. soln. |
| Solubility Information | Soluble in water 10mg/mL at 20°C. |
| Formula Weight | 40 |
| Odor | Odorless |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Sodium hydroxide |
Sécurité et manipulation
| Hazard Category | H290-H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only