missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.01 N Aqueous Solution, Spectrum™ Chemical

$118.38 - $413.53

Chemical Identifiers

CAS 1310-73-2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613517
View Documents
Spectrum Chemical Mfg Cor
S320500ML
500 mL Poly Bottle
Each for $118.38
Only null left
 
18613518
View Documents
Spectrum Chemical Mfg Cor
S3201LT
1 L Poly Bottle
Each for $196.06
Only null left
 
18613519
View Documents
Spectrum Chemical Mfg Cor
S3204LT
4 L Poly Bottle
Each for $316.16
Only null left
 
18613520
View Documents
Spectrum Chemical Mfg Cor
S32020LT
20 L Cubitainer
Each for $413.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-73-2

Specifications

1310-73-2 , 7732-18-5
500 mL
Chemical Solution
Sodium Hydroxide Solution
0.01 N ± 0.0002 N
5% ±0.25%
Poly Bottle
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.