missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.01 N Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S3201LT

 View more versions of this product

Catalog No. 18613518


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydroxide Solution
0.01 N ± 0.0002 N
5% ±0.25%
Poly Bottle
1310-73-2,7732-18-5
1 L
Chemical Solution

Chemical Identifiers

1310-73-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.