missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 6.0
Description
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
Solubility
Fully miscible in water.
Notes
Incompatible with heat and oxidizing agents.
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
Solubility
Fully miscible in water.
Notes
Incompatible with heat and oxidizing agents.
Spécifications
Spécifications
| Chemical Name or Material | Sodium citrate buffer soln. |
| Color | Colorless |
| Concentration | 0.5 M |
| pH | 6.0 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Odor | Odorless |
| MDL Number | MFCD00012462 |
| Solubility Information | Fully miscible in water. |
RUO – Research Use Only