missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, 10% (w/v), Ricca Chemical
$76.31 - $651.27
Chemical Identifiers
| CAS | 497-19-8 |
|---|---|
| Molecular Formula | CNa2O3 |
| Molecular Weight (g/mol) | 105.99 |
| MDL Number | MFCD00003494 |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| PubChem CID | 10340 |
| ChEBI | CHEBI:29377 |
| IUPAC Name | disodium carbonate |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7160-16
|
Ricca Chemical Company
716016 |
500 mL | Natural Poly Bottle |
Each for $76.31
|
|
||||
|
7160-32
|
Ricca Chemical Company
716032 |
1 L | Natural Poly Bottle |
Each for $105.56
|
|
||||
|
7160-1
|
Ricca Chemical Company
RI71601 |
4 L | Natural Poly Bottle |
Each for $215.34
|
|
||||
|
7160-25
|
Ricca Chemical Company
716025 |
10 L | Cubitainer |
Each for $651.27
|
|
||||
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| disodium carbonate |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 | |
| [Na+].[Na+].[O-]C([O-])=O |
Specifications
| 497-19-8,7732-18-5 | |
| 93.0,9.28 | |
| MFCD00003494 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium carbonate | |
| 10340 | |
| 10% | |
| 10% (w/v) | |
| Aqueous Solution | |
| 500 mL |
| 89.0,8.91 | |
| CNa2O3 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| Laboratory | |
| Natural Poly Bottle | |
| >12 | |
| Sodium Carbonate, 10% (w/v) |