missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, 10% (w/v), Ricca Chemical

Catalog No. 716016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
716016 500 mL Natural Poly Bottle
716032 1 L Natural Poly Bottle
71601 4 L Natural Poly Bottle
716025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 716016 Supplier Ricca Chemical Company Supplier No. 716016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH >12
Quantity 500 mL
Chemical Name or Material Sodium Carbonate, 10% (w/v)