missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 716025

 View more versions of this product

Catalog No. 7160-25


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Carbonate
497-19-8 , 7732-18-5
93.0,9.28
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
10340
10%
10% (w/v)
Aqueous Solution
10 L
10% (w/v)
89.0,8.91
CNa2O3
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
Laboratory
Cubitainer
>12

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium carbonate
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions