accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Electrochemistry Buffers
LC229622
Print
Sodium Borate Buffer Solution, LabChem™
Supplier:
LabChem
LC229622
Catalog No.
LC229622
Sign In
or
Register
to check your price and availability.
$100.29
/
Each
Quantity
Only null left
Explore available promotions
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Sodium Borate Buffer Solution
Color
Colorless
Decomposition Information
Boron
Quantity
1 L
Linear Formula
Na
2
B
4
O
7
·10H
2
O
Solubility Information
Soluble in water
CAS
7732-18-5
,
1303-96-4
,
1310-73-2
CAS Max %
99.74,0.25,0.01
Physical Form
Liquid
Identification
Passes Test
Packaging
Poly Bottle
Formula Weight
381.37
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions