missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, Saturated, for Neutralization, Certified, LabChem™
Supplier: LabChem LC229474
Specifications
| Sodium Bicarbonate | |
| Saturated, for Neutralization | |
| 90.4,9.6 | |
| NaHCO3 | |
| 1.1g/mL | |
| Soluble in water | |
| [Na+].OC([O-])=O | |
| 84.01 | |
| CHEBI:32139 | |
| Certified | |
| Poly Bottle | |
| Fume; Carbon monoxide; Carbon dioxide | |
| Liquid |
| Colorless | |
| 144-55-8 , 7732-18-5 | |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| sodium hydrogen carbonate | |
| 516892 | |
| 84.01 | |
| Passes Test | |
| 1.1g/mL | |
| 4 L |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| 516892 | |
| sodium hydrogen carbonate |
| CHNaO3 | |
| MFCD00003528 | |
| sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature