missing translation for 'onlineSavingsMsg'
Learn More

Sodium AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Na), LabChem™

$54.63 - $98.40

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC228107
View Documents
LabChem
LC228107
125 mL Poly Bottle N/A
Only null left
 
LC228101
View Documents
LabChem
LC228101
500 mL Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride

Specifications

Poly Bottle
99.75,0.25
1.01cSt
ClNa
NaCl
Colorless
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
125 mL
CHEBI:26710
Certified
7647-14-5,7732-18-5
Passes Test
Liquid
1000 ppm ±5 ppm (1mL = 1mg Na)
MFCD00003477
Soluble in water
[Na+].[Cl-]
58.44
5234
58.44
Sodium AA Standard, 1000ppm (1mL = 1mg Na)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'