missing translation for 'onlineSavingsMsg'
Learn More

Sodium AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Na), LabChem™

Supplier:  LabChem LC228107

 View more versions of this product

Catalog No. LC228107


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium AA Standard
Poly Bottle
99.75,0.25
1.01cSt
ClNa
NaCl
Colorless
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
125 mL
CHEBI:26710
Certified
1000ppm (1mL = 1mg Na)
7647-14-5,7732-18-5
Passes Test
Liquid
1000 ppm ±5 ppm (1mL = 1mg Na)
MFCD00003477
Soluble in water
[Na+].[Cl-]
58.44
5234
58.44

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions