missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Na), LabChem™
Supplier: LabChem LC228101
Specifications
| Sodium AA Standard | |
| Poly Bottle | |
| 99.75,0.25 | |
| 1.01cSt | |
| ClNa | |
| NaCl | |
| Colorless | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 | |
| Certified |
| 1000ppm (1mL = 1mg Na) | |
| 7647-14-5,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±5 ppm | |
| MFCD00003477 | |
| Soluble in water | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 | |
| 58.44 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature