Learn More
Eisco™ Single and Tandem Pulleys
Single pulley plus double and triple tandem pulleys in durable nickel/steel frames.
Supplier:  Eisco™ PH0281A
 Afficher plus de versions de ce produit
Numéro de catalogue. S96181
Il en reste null
Description
Description
  • Plastic pulleys mounted on brass bearings and set in nickel/steel frames with hooks at the top and the bottom
  • Single pulley measures 2 in. (5cm) Double tandem pulley block, measuring 5.75 in. (14.6cm), holds two plastic pulleys with successive diameters of 1.52 and 2 in. (3.8 and 5cm)
  • Triple tandem pulley block, measuring 7.18 in. (18cm), holds three plastic pulleys with successive diameters of 1, 1.52 and 2 in. (2.5, 3.8 and 5cm)
Spécifications
Identifiants chimiques
Plastic, Nickel/Steel
Open sided frame, hooks
Spécifications
Single Tandem Pulley
9.84 in.
1.96 in.
Open sided frame, hooks
44 cm
2.2 lb.
6.29 in.
Plastic, Nickel/Steel
25 cm
5 cm
17.32 in.
1 kg
16 cm
44 x 16 x 5 cm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS