missing translation for 'onlineSavingsMsg'
Learn More

First Safety™ Single Cylinder Hand Trucks

For safe, convenient transport of medical, industrial, and commercial use gases.

Supplier:  First Safety™ 35004

 Afficher plus de versions de ce produit

Numéro de catalogue. 19063135


Il en reste null

Description

Description

  • Four wheel cylinder industrial quality hand truck for gas cylinders up to 12.0 inch diameter
  • Designed and built in the USA using American steel, labor, assembly, packaging and finishes
  • Configured for one cylinder with 8 in. semi-pneumatic wheels
  • Swing-down steel carriages allow use of rear swivel casters for additional safety and control
  • Fold flat against rear of hand truck for storage in the upright position
  • Upper handle detaches for shipment
  • Exterior grade powder coated for durability
  • Constructed from heavy 11 gauge HR Steel with 3/16 inch thick steel supports and round steel tube
  • Standard 1.5 inch poly strap with cinch buckle system, lab tested to 1138 PSI
  • 700 lb. capacity
  • Units are fully welded
Spécifications

Identifiants chimiques

Hand truck
HR Steel (11 ga.)
47 in.
1 Cylinder up to 12 in. dia.
Safe, convenient transport of medical, industrial, and commercial use gases
Poly straps with steel cinch buckles

Spécifications

Hand truck
700 lb.
Main wheels: Semi pneumatic; 8 in. diameter
Safe, convenient transport of medical, industrial, and commercial use gases
Poly straps with steel cinch buckles
19 in.
1 Cylinder up to 12 in. dia.
HR Steel (11 ga.)
19 x 19 x 47 in.
47 in.
44 lb.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions