missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0250 N (N/40), Ricca Chemical
ASTM for Chloride Testing
$190.33 - $456.13
Chemical Identifiers
| Molecular Formula | AgNO3 |
|---|---|
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
Chemical Identifiers
| AgNO3 | |
| MFCD00003414 | |
| silver(1+) nitrate |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| 7732-18-5 | |
| MFCD00003414 | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| Amber Poly Bottle | |
| ∼6 | |
| Silver Nitrate |
| AgNO3 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| Laboratory | |
| ASTM for Chloride | |
| 1 L |