missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.0250 N (N/40), Ricca Chemical

ASTM for Chloride Testing

Supplier:  Ricca Chemical Company 692032

 View more versions of this product

Catalog No. 6920-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
AgNO3
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
Laboratory
ASTM for Chloride
1 L
7732-18-5
MFCD00003414
[Ag+].[O-][N+]([O-])=O
169.87
Amber Poly Bottle
∼6

Chemical Identifiers

AgNO3
MFCD00003414
silver(1+) nitrate
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions