missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0250 N (N/40), Ricca Chemical
ASTM for Chloride Testing
Supplier: Ricca Chemical Company 69201
Specifications
| Silver Nitrate | |
| AgNO3 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| Laboratory | |
| ASTM for Chloride | |
| 4 L |
| 7732-18-5 | |
| MFCD00003414 | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| Black Poly Bottle | |
| ∼6 |
Chemical Identifiers
| AgNO3 | |
| MFCD00003414 | |
| silver(1+) nitrate |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| [Ag+].[O-][N+]([O-])=O |