missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.0192 N (0.0192 M), 1 mL = 1 mg CN-, Ricca Chemical
$106.53 - $521.23
Chemical Identifiers
| Molecular Formula | AgNO3 |
|---|---|
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| IUPAC Name | silver(1+) nitrate |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
691016
|
Ricca Chemical Company
691016 |
500 mL | Amber Poly Bottle |
Each for $106.53
|
|
||||
|
6910-32
|
Ricca Chemical Company
691032 |
1 L | Amber Poly Bottle |
Each for $187.33
|
|
||||
|
6910-1
|
Ricca Chemical Company
69101 |
4 L | Black Poly Bottle |
Each for $521.23
|
|
||||
Chemical Identifiers
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
Specifications
| Colorless | |
| 98.0,0.32 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| 24470 | |
| Laboratory | |
| ∼6 | |
| Liquid | |
| Silver Nitrate |