missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.0192 N (0.0192 M), 1 mL = 1 mg CN-, Ricca Chemical

Supplier:  Ricca Chemical Company 691032

 View more versions of this product

Catalog No. 6910-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
7761-88-8 , 7732-18-5
100.0,0.34
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
1 mL = 1 mg CN
CHEBI:32130
Amber Poly Bottle
1 L
0.0192Normal,0.0192Molar
Colorless
98.0,0.32
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
24470
Laboratory
∼6
Liquid

Chemical Identifiers

AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions