Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Catalog No. 290MN211
Change view
Click to view available options
Type:
Capillary Column Connector
Replacement Column Connector Nuts
Diameter (Metric) Inner:
0.1 to 0.25 mm
0.1 to 0.53 mm
0.45 to 0.53 mm
32 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Diameter (Metric) Inner
290MN211 Replacement Column Connector Nuts 0.1 to 0.53 mm
03150326 Capillary Column Connector 0.1 to 0.25 mm
03150327 Capillary Column Connector 32 mm
03150328 Capillary Column Connector 0.45 to 0.53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 290MN211 Supplier Thermo Scientific™ Supplier No. 290MN211
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
Specifications
Type Replacement Column Connector Nuts
Description 290 MN211
Diameter (Metric) Inner 0.1 to 0.53 mm
For Use With (Equipment) Fused Silica Capillary Column
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.