missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ N-DEX™ 6005PF Disposable Nitrile Gloves

Catalog No. 29635199
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29635199 Small
29635200 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29635199 Supplier SHOWA™ Supplier No. 6005PFS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-DEX™ 6005 series powder free gloves provide enhanced dexterity and performance across a range of tactical applications including food, electronics, janitorial, municipal services and utilities.

  • Easy to put on and remove
  • Can easily be one-hand used
  • Ambidextrous
  • Rolled cuff
  • Smooth grip
  • Made in the U.S.A.
  • Specific applications include food handling, agriculture, washing and cleaning, public utility and coroner/morgue

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 455, Cat. I
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6005PFSM
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less