missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ SeQuant™ ZIC™-HILIC (3.5μm, 200Å) Capillary HPLC Column
Description
- Stationary Phase: Zwitterionic functional groups (1:1 charge balance) covalently bonded to porous silica
- Pore Size: 200 Å
- Applications: Separation of polar and hydrophilic compounds with orthogonal selectivity to reversed-phase chromatography
- Compatibility: Compatible with a range of buffers, organic solvents, and temperatures for MS, ELSD, and UV detection techniques
- Performance Characteristics: High reproducibility, robust bonded phase, and pH-independent stability
Specifications
Specifications
| Diameter (Metric) | 0.3 mm |
| Length (Metric) | 30 mm |
| Mobile Phase Compatibility | HILIC (hydrophilic interaction) |
| Particle Size | 3.5 μm |
| Pore Size | 200 Å |
| Stationary Phase | Zwitterionic (ZIC) |
| Product Line | ZIC-HILIC |
| Column Type | SeQuant™ Capillary HPLC Column |
| Phase | HILIC |
| Material | PEEK |
| Show More |