Learn More
Cytiva Sepharose Agarose Gel Filtration Media
Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.
Supplier: Cytiva 17011001
Description
European Pharmacopoeia Solutions
- Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
- All are manufactured and tested in compliance with the Ph. Eur.
- Supplied with a Certificates of Analysis
- Lot No. and expiry date are stated on all product labels
Specifications
| 1 L | |
| 45 to 165 μm | |
| Solid Bead | |
| Allow characterization or cleaning up of samples containing components of diverse molecular weight | |
| 10cm | |
| 4 to 9 | |
| 4 to 30°C, 20% Ethanol |
| <14 cm/hr. | |
| Sepharose Agarose Gel Filtration Media | |
| 5cm | |
| 3.94 in. | |
| 100-200 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, 5 cm i.d. | |
| 6% Agarose |