missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sepharose Agarose Gel Filtration Media

Catalog No. p-3676373
Change view
Click to view available options
Flow Rate:
<11.5 cm/hr.
<14 cm/hr.
<26 cm/hr.
<30 cm/hr.
Particle Size:
45 to 165 μm
pH Range:
3 to 12
3 to 13
4 to 9
Support Type:
2% Agarose
4% Agarose
6% Agarose
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Particle Size pH Range Support Type
45000032 <26 cm/hr. 45 to 165 μm 3 to 13 2% Agarose
45000029 <11.5 cm/hr. 45 to 165 μm 4 to 9 4% Agarose
45000033 <30 cm/hr. 45 to 165 μm 3 to 12 6% Agarose
45000028 <14 cm/hr. 45 to 165 μm 4 to 9 6% Agarose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45000032 Supplier Cytiva Supplier No. 17015001
Only null left

Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.

European Pharmacopoeia Solutions

  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels

Order Info

Supplied in 1L plastic containers

Specifications

Quantity 1 L
Flow Rate <26 cm/hr.
Particle Size 45 to 165 μm
Product Type Sepharose Agarose Gel Filtration Media
Format Solid Bead
Diameter (Metric) Inner 5cm
For Use With (Application) Allow characterization or cleaning up of samples containing components of diverse molecular weight
Height (English) 3.94 in.
Height (Metric) 10cm
Max. Pressure 80-150 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, column 5 cm i.d.
pH Range 3 to 13
Support Type 2% Agarose
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less