missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G75 Superfine Media

Catalog No. 45000024
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000024 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000024 Supplier Cytiva Supplier No. 17005101

Classic gel filtration medium

  • Fractionation range, globular proteins: 3000 to 70,000
  • Particle size, wet: 20 to 155μm
  • Working pH range: 2-10

Specifications

Quantity 100 g
Flow Rate 1.5 mL/min.
Particle Size 22 to 143 μm in wet form, 10 to 40 μm in dry form
Bed Volume 12 to 15mg/g
Product Type Sephadex G75 Media
Format Solid Bead
For Use With (Application) Desalting and Buffer exchange of large biomolecules, for specific cases requiring very high resolution
Max. Pressure Care must be taken to avoid compressing the gel. Do not increase the pressure beyond 0.16 bar.
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type DNA
Content And Storage 4° to 30°C
Show More Show Less