missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G75 Media

Catalog No. 45000022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000022 Supplier Cytiva Supplier No. 17005001

Suitable for gel filtration

  • For purposes of determining maximum flowrate, beads behave as rigid spheres according to Darcy's Law
  • Maximum operating pressure 0.15 bar
  • Working pH range 2 to 10
  • Particle size range of wet beads 90 to 280μm
  • Bed volume in distilled water 12 to 15mL/g
  • Fractionation ranges: 1 x 103 to 5 x 104 for dextrans, 3 x 103 to 8 x 104 for globular proteins

Specifications

Quantity 100 g
Flow Rate < Darcy´s Law
Particle Size 90 to 280 μm in wet form, 40 to 120 μm in dry form
Bed Volume 12 to 15mg/g
Product Type Sephadex G75 Media
Format Solid Bead
Diameter (Metric) Inner 5cm
For Use With (Application) Desalting and Buffer exchange of large biomolecules >80000 M.W.
Height (English) 3.93 in.
Height (Metric) 10cm
Max. Pressure min 90 cm/h, bed height 10 cm, column 5 cm i.d.
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type DNA
Content And Storage 4 to 30°C
Show More Show Less