missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G15 Media

Catalog No. 45000013
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000013 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000013 Supplier Cytiva Supplier No. 17002001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gel filtration medium for desalting and buffer exchange for small biomolecules greater than 1500 molecular weight

  • For purposes of determining maximum flowrate and maximum pressure, beads behave as rigid spheres according to Darcy's Law
  • Working pH range 2 to 13
  • Particle size range for wet beads 60 to 180μm
  • Bed volume in distilled water 2.5 to 3.5mL/g
  • Exclusion limit: <1.5 x 103 for dextrans, <1.5 x 103 for globular proteins

Specifications

Quantity 100 g
Flow Rate < Darcy´s Law
Particle Size 60 to 180 μm in wet form, 40 to 120 μm in dry form
Bed Volume 2.5 to 3.5mL/g in Distilled Water
Product Type Sephadex G15 Media
Format Solid Bead
Diameter (Metric) Inner 2.6cm
For Use With (Application) Useful for desalting and Buffer exchange of Peptides and small biomolecules >1500 M.W.
Height (English) 11.81 in.
Height (Metric) 30cm
Max. Pressure 20-47 cm/h, pressure drop cm H2O/bed height = 2, bed height 30 cm, 2.6 cm i.d.
pH Range 2 to 13
Support Type Cross-linked Dextran
Sample Type Peptides and small biomolecules
Content And Storage 4 to 30°C
Show More Show Less