missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex G-50 M DNA Grade

Catalog No. 45000021
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000021 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000021 Supplier Cytiva Supplier No. 17004502
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sephadex G-50 M DNA Grade

Specifications

Quantity 100 g
Product Type Sephadex G50 Media
Format Solid Bead
For Use With (Application) Desalting and Buffer exchange of biomolecules
Support Type Cross-linked Dextron
Sample Type DNA
Content And Storage 4° to 30°C