missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G-100 Media

Catalog No. p-3676369
Change view
Click to view available options
Flow Rate:
1 mL/min.
4.2 mL/min.
Particle Size:
100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Particle Size
45000025 4.2 mL/min. 100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
45000027 1 mL/min. 25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000025 Supplier Cytiva Supplier No. 17006001
Only null left

Commonly used for desalting and buffer exchange of very large biomolecules

  • Available in two particle size ranges

Fine particle size

  • Particle size, dry: 40 to 120μm
  • Fractionation range, globular proteins: 4000 to 150,000
Superfine particle size
  • Particle size, dry: 10 to 40μm
  • Fractionation range, globular proteins: 4000 to 100,000

Specifications

Quantity 100 g
Flow Rate 4.2 mL/min.
Particle Size 100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
Bed Volume 15 to 20mL/g in Distilled Water
Product Type Sephadex G-100 Media
Format Solid Bead
For Use With (Application) Useful for desalting and Buffer exchange of very large biomolecules
Max. Pressure 0.096 bar
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type Large biomolecules
Content And Storage 4° to 30°C
Show More Show Less