Learn More
Sensidyne™ Detector Tubes: Carbon Monoxide
Detector tubes
Supplier:  Sensidyne™ 106UH
 Afficher plus de versions de ce produit
Numéro de catalogue. 18068F
Il en reste null
Description
Description
All detector tubes are sold in packs of 10.
Spécifications
Identifiants chimiques
Carbon Monoxide
10
Spécifications
Carbon Monoxide
0.5/1
2 min.
0°C to 40°C
0.1 to 20%
10
32°F to 104°F
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS