missing translation for 'onlineSavingsMsg'
Learn More

Selenium ICP Standard, 10,000 ppm Se in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PSE10KN100

Packaging Natural LDPE Bottle

 View more versions of this product

Catalog No. PSE10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7782-49-2 , 7697-37-2 , 7732-18-5
78.971
6326970
selenium
Se
BUGBHKTXTAQXES-UHFFFAOYSA-N
CHEBI:27568
[Se]

Specifications

Selenium ICP Standard
7782-49-2 , 7697-37-2 , 7732-18-5
86.0, 11.57, 0.86
1 mL = 10 mg Se
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
Natural LDPE Bottle
10,000ppm Se
89.0, 12.05, 0.89
Se
[Se]
78.971
CHEBI:27568
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents