missing translation for 'onlineSavingsMsg'
Learn More

Selenium ICP-MS Standard, 1000 ppm Se in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company MSSE1KN100

Catalog No. MSSE1KN-100

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Selenium ICP-MS Standard
7782-49-2,7697-37-2,7732-18-5
93.0,4.76,0.09
1 mL = 1 mg Se
Se
Colorless
Approximately 0°C
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
100 mL
CHEBI:27568
Natural LDPE Bottle
1,000ppm Se
97.0,4.95,0.1
Liquid
MFCD00134090 MFCD00011224
< 2
Approximately 100°C
Miscible with water
ICP Single-element Standard Solution
[Se]
78.97
6326970
Laboratory

Chemical Identifiers

7782-49-2
78.97
BUGBHKTXTAQXES-UHFFFAOYSA-N
CHEBI:27568
[Se]
Se
MFCD00134090 MFCD00011224
6326970
selenium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions