missing translation for 'onlineSavingsMsg'
Learn More

3M™ Secure Click™ Half Facepiece Reusable Respirator

Catalog No. 19072214
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19072214 Small
19072216 Medium
19072213 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19072214 Supplier 3M™ Supplier No. 7100172045

Secure Click™ style enables a quick and simple connection of filters and cartridges. Half-face respirator also comes equipped with speaking diaphragm for easy communication.

  • Designed with smart, and intuitive features, the HF-800 Series is simple, comfortable, and reliable
  • The quick, and easy one-touch negative pressure push button seal check provides improved confidence in respirator fit
  • Secure Click™ connection style enables a quick, simple connection of filters and/or cartridges
  • Dual-flow cartridges allow for easier breathing and greater comfort
  • Patented silicone flex-joint faceseal provides a soft feel
  • Easy-adjust head cradle buckles provide two-way adjustability

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Secure Click™
Color Gray
Material Silicone
Product Type Half-Mask Respirator
Size Small
Facepiece Material Silicone
For Use With (Application) Sanding, Masonary, Chemical Clean-up, Laboratories, Grinding, Sawing, Machining, Cleaning, Painting, Chiseling, Assembly, Furnace Operations, Chipping, Pouring and Casting
For Use With (Equipment) 3M Secure Click™ Cartridges and Filters
Includes Speaking Diaphragm
Model HF-801SD
Width (Metric) 125 mm
Height (Metric) 134 mm
Show More Show Less