missing translation for 'onlineSavingsMsg'
Learn More

Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek

Ideal for peak identification work

$1,310.92 - $3,771.53

Specifications

Certifications/Compliance DOT 39 NRC
Diameter (Metric) 10.2 cm
Diameter (English) 4 in.
Volume (Metric) Gas 48 L
For Use With (Application) Ideal for peak identification work
View More Specs

Products 9
Catalog Number Mfr. No. Concentration Components Type Shelf Life Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Components Type Shelf Life Price Quantity & Availability  
06802417
Encompass
View Documents Promotion Details
N/A 99.80% Carbon Dioxide Air Standard 2 Years
Each for $1,310.92
Only null left
 
06802470
Encompass
View Documents Promotion Details
N/A CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard 2 Years
Each for $2,663.79
Only null left
 
06802476
Encompass
View Documents Promotion Details
N/A CO 1%, CO2 1%, Methane 1%, Ethane 1%, Ethylene 1%, Ecetylene 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene, Ecetylene in Nitrogen Transportable Air Standard 2 Years
Each for $2,451.06
Only null left
 
06802473
Encompass
View Documents Promotion Details
N/A CO 1%, CO2 1%, H2 1%, O2 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard 2 Years
Each for $3,195.38
Only null left
 
06802490
Encompass
View Documents Promotion Details
N/A 1000 ppm C1-C6 n-Paraffins in Helium Transportable Air Standard 2 Years
Each for $3,771.53
Only null left
 
06802493
Encompass
View Documents Promotion Details
N/A 100 ppm C1-C6 n-Paraffins in Nitrogen Transportable Air Standard 2 Years
Each for $2,905.28
Only null left
 
06802497
Encompass
View Documents Promotion Details
N/A 100 ppm C2-C6 Olefins in Nitrogen Transportable Air Standard 2 Years
Each for $3,195.38
Only null left
 
06802504
Encompass
View Documents Promotion Details
N/A 15 ppm n-Butane, Isobutane, cis-2-Butene, trans-2-Butene, 1-Butene, Isobutene, 1,3-Butadiene, Ethyl Acetylene in Nitrogen Transportable Air Standard 1 Year
Each for $2,643.98
Only null left
 
06802502
Encompass
View Documents Promotion Details
N/A 15 ppm Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, n-Butane, Propyne in Nitrogen Transportable Air Standard 1 Year
Each for $2,956.23
Only null left
 
Description

Description

  • CGA 165 connection and can deliver large volumes of sample
  • Scotty™ (Air Liquide) 48
  • Contents: 48L
  • Pressure: 300 psig (21 bar)
  • Outlet Fitting: CGA 165
  • Weight: 1.75 lb. (0.8kg)
  • Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
  • DOT Specifications: 39 NRC

All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements

Specifications

Specifications

DOT 39 NRC
4 in.
Ideal for peak identification work
41 cm
1.75 lb.
0.8 kg
10.2 cm
48 L
16.25 in.
300 psi
12.68 gal.
Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 Years

SDS
Documents

Documents