missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
Description
- CGA 165 connection and can deliver large volumes of sample
- Scotty™ (Air Liquide) 48
- Contents: 48L
- Pressure: 300 psig (21 bar)
- Outlet Fitting: CGA 165
- Weight: 1.75 lb. (0.8kg)
- Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
- DOT Specifications: 39 NRC
All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements
Specifications
Specifications
| Certifications/Compliance | DOT 39 NRC |
| Components | C2-C6 Olefins in Nitrogen |
| Diameter (Metric) | 10.2 cm |
| Diameter (English) | 4 in. |
| Concentration | 100 ppm |
| Volume (Metric) Gas | 48 L |
| For Use With (Application) | Ideal for peak identification work |
| Height (English) | 16.25 in. |
| Height (Metric) | 41 cm |
| Pressure | 300 psi |
| Show More |