missing translation for 'onlineSavingsMsg'
Learn More

Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek

Ideal for peak identification work

Supplier:  Chromatographic RK 34505

Encompass

 View more versions of this product

Catalog No. 06802470


Only null left

Description

Description

  • CGA 165 connection and can deliver large volumes of sample
  • Scotty™ (Air Liquide) 48
  • Contents: 48L
  • Pressure: 300 psig (21 bar)
  • Outlet Fitting: CGA 165
  • Weight: 1.75 lb. (0.8kg)
  • Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
  • DOT Specifications: 39 NRC

All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements

Specifications

Chemical Identifiers

DOT 39 NRC
4 in.
Ideal for peak identification work
41 cm
1.75 lb.
0.8 kg
10.2 cm
48 L
16.25 in.
300 psi
12.68 gal.

Specifications

DOT 39 NRC
10.2 cm
CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen
Ideal for peak identification work
41 cm
1.75 lb.
0.8 kg
Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen
4 in.
48 L
16.25 in.
300 psi
12.68 gal.
Transportable Air Standard
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 Years

SDS
Documents

Documents

Promotions

Promotions