missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLSC22X

 View more versions of this product

Catalog No. PLSC2-2X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Scandium Standard
12060-08-1 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00011223
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
2% HNO3
137.91
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,2.0,97.9
O3Sc2
AA, ICP
Certified Reference Material
[O--].[O--].[O--].[Sc+3].[Sc+3]
discandium(3+) trioxidandiide
500 mL
Matrix    Concentration    Cat. No.2-5% HNO3    1000μg/mL    PLSC2-2Y, -2X    10,000    PLSC2-3Y, -3X

Chemical Identifiers

12060-08-1
137.91
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
[O--].[O--].[O--].[Sc+3].[Sc+3]
O3Sc2
MFCD00011223
discandium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only