CAS RN 12060-08-1
Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium ICP Standard, 10,000 ppm Sc in 5% HNO3, Ricca Chemical
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium AA Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical
CAS: 12060-08-1
Scandium ICP-MS Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
ICP-MS Internal Standard, 10 ppm Sc in 3% HNO3, Ricca Chemical
CAS: 12060-08-1
Scandium ICP Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium Standard For ICP, 1000μg/mL in 2% HNO3, SPEX CertiPrep™
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium Standard For ICP/MS, SPEX CertiPrep™
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]
Scandium Standard For ICP, 1000μg/mL, SPEX CertiPrep™
CAS: 12060-08-1 Molecular Formula: O3Sc2 Molecular Weight (g/mol): 137.91 MDL Number: MFCD00011223 InChI Key: HJGMWXTVGKLUAQ-UHFFFAOYSA-N IUPAC Name: discandium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Sc+3].[Sc+3]