missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLSC23X

 View more versions of this product

Catalog No. PLSC2-3X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Scandium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011223
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
5% HNO3
137.91
Assurance
Triple-leached LDPE bottle
12060-08-1 , 7697-37-2 , 7732-18-5
O3Sc2
AA, ICP
Certified Reference Material
[O--].[O--].[O--].[Sc+3].[Sc+3]
discandium(3+) trioxidandiide
500 mL

Chemical Identifiers

12060-08-1
137.91
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
[O--].[O--].[O--].[Sc+3].[Sc+3]
O3Sc2
MFCD00011223
discandium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only