missing translation for 'onlineSavingsMsg'
Learn More

Scandium ICP Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical

Catalog No. PSC1KN100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PSC1KN100 100 mL Natural LDPE Bottle
PSC1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PSC1KN100 Supplier Ricca Chemical Company Supplier No. PSC1KN100

Chemical Identifiers

CAS 12060-08-1 , 7697-37-2 , 7732-18-5
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sc
CAS Min % 93.0, 5.34, 0.14
CAS Max % 96.0, 5.56, 0.15
Equivalence 1 mL = 1 mg Sc
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Scandium ICP Standard
Show More Show Less