missing translation for 'onlineSavingsMsg'
Learn More

Scandium ICP Standard, 10,000 ppm Sc in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PSC10KN100

Catalog No. PSC10KN-100


Only null left

Description

Description

Specifications

Specifications

Scandium ICP Standard
12060-08-1 , 7697-37-2 , 7732-18-5
81.0,16.22,1.24
1 mL = 10 mg Sc
O3Sc2
Colorless
Approximately 100°C
Miscible with water
Standard
[O--].[O--].[O--].[Sc+3].[Sc+3]
137.91
Laboratory
Natural LDPE Bottle
10,000ppm Sc
84.0,16.88,1.29
Liquid
MFCD00011223
< 2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
discandium(3+) trioxidandiide
100 mL

Chemical Identifiers

12060-08-1
137.91
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
[O--].[O--].[O--].[Sc+3].[Sc+3]
O3Sc2
MFCD00011223
discandium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions