missing translation for 'onlineSavingsMsg'
Learn More

Scandium ICP-MS Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical

Catalog No. MSSC1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSSC1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSSC1KN100 Supplier Ricca Chemical Company Supplier No. MSSC1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12060-08-1 , 7697-37-2 , 7732-18-5
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sc
CAS Min % 93.0, 5.34, 0.14
CAS Max % 96.0, 5.56, 0.15
Equivalence 1 mL = 1 mg Sc
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Scandium ICP-MS Standard
Show More Show Less