missing translation for 'onlineSavingsMsg'
Learn More

Samarium ICP Standard, 1000 ppm Sm in 3% HNO3, Ricca Chemical

Catalog No. PSM1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSM1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSM1KN100 Supplier Ricca Chemical Company Supplier No. PSM1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12060-58-1 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sm
CAS Min % 94.0, 4.36, 0.11
CAS Max % 97.0, 4.53, 0.11
Equivalence 1 mL = 1 mg Sm
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Samarium ICP Standard
Show More Show Less